4-Aminopyrimidine, Sigma-Aldrich®
Supplier: Merck
Warning
M-Clarity™ quality level = MQ100
261823-1G 261823-250MG
SIAL261823-1GEA 98.9 EUR
SIAL261823-1G SIAL261823-250MG
4-Aminopyrimidine, Sigma-Aldrich®
4-Aminopyrimidine
Formula: C₄H₅N₃
MW: 95.1 g/mol
Melting Pt: 154…156 °C
MDL Number: MFCD00006112
CAS Number: 591-54-8