2,4,6-Trimethyl-m-phenylenediamine, Sigma-Aldrich®

Supplier: Merck


M-Clarity™ quality level = MQ200

263885-100G 263885-25G
SIAL263885-100GEA 416 EUR
SIAL263885-100G SIAL263885-25G
2,4,6-Trimethyl-m-phenylenediamine, Sigma-Aldrich®
2,4,6-Trimethyl-m-phenylenediamine
Formula: C₉H₁₄N₂
MDL Number: MFCD00010149
CAS Number: 3102-70-3
EINECS: 221-456-9
UN: 2811
ADR: 6.1,I

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR