ω-Chloro-2,4-difluoroacetophenone, Sigma-Aldrich®
Supplier: Merck
Synonyms: 2-Chloro-1-(2,4-difluorophenyl)ethanone, α-Chloro-2,4-difluoroacetophenone, 2-Chloro-2',4'-difluoroacetophenone
M-Clarity™ quality level = MQ200
272507-25G 272507-5G
SIAL272507-25GEA 112 EUR
SIAL272507-25G SIAL272507-5G
ω-Chloro-2,4-difluoroacetophenone, Sigma-Aldrich®
ω-Chloro-2,4-difluoroacetophenone
Formula: C₈H₅ClF₂O
Boiling Pt: 233 °C (1013 hPa)
Melting Pt: 47…48 °C
MDL Number: MFCD00013252
CAS Number: 51336-94-8
UN: 3261
ADR: 8,II