Dysprosium trinitrate hydrate, Sigma-Aldrich®

Supplier: Merck

Synonyms: Dysprosium (III) nitrate hydrate

M-Clarity™ quality level = MQ100

298158-100G 298158-25G
SIAL298158-100GEA 231 EUR
SIAL298158-100G SIAL298158-25G
Dysprosium trinitrate hydrate, Sigma-Aldrich®
Dysprosium trinitrate hydrate
Formula: DyN₃O₉·H₂O
CAS Number: 100641-13-2
EINECS: 233-410-5
UN: 1477
ADR: 5.1,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR