Bis(2-chloroethyl)ether, Sigma-Aldrich®
Supplier: Merck
Synonyms: 2-Chloroethyl ether
M-Clarity™ quality level = MQ200
35660-1L 35660-250ML 35660-50ML
SIAL35660-1LEA 154 EUR
SIAL35660-1L SIAL35660-250ML SIAL35660-50ML
Bis(2-chloroethyl)ether, Sigma-Aldrich®
Bis(2-chloroethyl) ether
Formula: C₄H₈Cl₂O
Boiling Pt: 178 °C (1013 hPa)
Melting Pt: –44 °C
Density: 1.22 g/cm³ (20 °C)
Flash Pt: 55 °C
MDL Number: MFCD00000975
CAS Number: 111-44-4
EINECS: 203-870-1
UN: 1916
ADR: 6.1,II
Merck Index: 13,03091