Hept-6-enoic acid, Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ100
358940-1ML
358940-5ML
SIAL358940-1MLEA
133
EUR
SIAL358940-1ML
SIAL358940-5ML
Hept-6-enoic acid, Sigma-Aldrich®
Hept-6-enoic acid
Formula:
C₇H₁₂O₂ Boiling Pt: 70…72 °C (0.5 mmHg) Melting Pt: –6.5 °C Density: 0.9515 g/cm³ (14.9 °C) |
MDL Number:
MFCD00014382 CAS Number: 1119-60-4 EINECS: 214-283-5 UN: 3265 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...