2-Chloro-5-(trifluoromethyl)pyridine, Sigma-Aldrich®
Supplier: Merck
Synonyms: 2-Chloro-5-trifluoromethylpyridine
M-Clarity™ quality level = MQ100
366137-10G 366137-50G
SIAL366137-10GEA 39 EUR
SIAL366137-10G SIAL366137-50G
2-Chloro-5-(trifluoromethyl)pyridine, Sigma-Aldrich®
2-Chloro-5-(trifluoromethyl)pyridine
Formula: C₆H₃ClF₃N
Boiling Pt: 152 °C (1013 hPa)
Melting Pt: 30…31 °C
Density: 1.41 g/cm³ (20 °C)
MDL Number: MFCD00042225
CAS Number: 52334-81-3
EINECS: 257-856-5
UN: 2811
ADR: 6.1,III