2-Amino-5-methylthiazole, Sigma-Aldrich®
Supplier: Merck
Synonyms: 5-Methylthiazol-2-ylamine
M-Clarity™ quality level = MQ100
380563-25G 380563-5G
SIAL380563-25GEA 62.7 EUR
SIAL380563-25G SIAL380563-5G
2-Amino-5-methylthiazole, Sigma-Aldrich®
2-Amino-5-methylthiazole
Formula: C₄H₆N₂S
MW: 114.17 g/mol
Boiling Pt: 222
Melting Pt: 96
MDL Number: MFCD00078317
CAS Number: 7305-71-7
EINECS: 423-800-5