1,3-Dimethylurea, Sigma-Aldrich®
Supplier: Merck
Synonyms: N,N'-Dimethylurea
40410-1KG 40410-50G
SIAL40410-1KGEA 47.2 EUR
SIAL40410-1KG SIAL40410-50G
1,3-Dimethylurea, Sigma-Aldrich®
1,3-Dimethylurea
Formula: C₃H₈N₂O
Boiling Pt: 268 °C (1013 hPa)
Melting Pt: 96 °C
Density: 1.142 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00008286
CAS Number: 96-31-1
EINECS: 202-498-7