Allyltrimethoxysilane, Sigma-Aldrich®

Supplier: Merck

Synonyms: Trimethoxyallylsilane

M-Clarity™ quality level = MQ100

446955-25ML 446955-5ML
SIAL446955-25MLEA 143 EUR
SIAL446955-25ML SIAL446955-5ML
Allyltrimethoxysilane, Sigma-Aldrich®
Allyltrimethoxysilane
Formula: H₂CCHCH₂Si(OCH₃)₃
MW: 162.26 g/mol
MDL Number: MFCD00053865
CAS Number: 2551-83-9
EINECS: 219-855-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR