2,2'-Methyliminodiethanol, Sigma-Aldrich®
Supplier: Merck
Warning
Synonyms: N,N-Diethanolmethylamine, N-Methyldiethanolamine, N-Methyl-2,2'-iminodiethanol, AMIETOL M 12, MDEA
M-Clarity™ quality level = MQ200
471828-25ML 471828-18L 471828-250ML 471828-2L
SIAL471828-25MLEA 28.4 EUR
SIAL471828-25ML SIAL471828-18L SIAL471828-250ML SIAL471828-2L
2,2'-Methyliminodiethanol, Sigma-Aldrich®
2,2'-Methyliminodiethanol
Formula: H₃CN(CH₂CH₂OH)₂
MW: 119.16 g/mol
Boiling Pt: 247
Melting Pt: –21 °C
Density: 1.04 g/cm³ (20 °C)
Flash Pt: 137
MDL Number: MFCD00002848
CAS Number: 105-59-9
EINECS: 203-312-7