4-Bromo-m-toluonitrile, Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ100
566365-25G 566365-5G
SIAL566365-25GEA 282 EUR
SIAL566365-25G SIAL566365-5G
4-Bromo-m-toluonitrile, Sigma-Aldrich®
4-Bromo-m-toluonitrile
Formula: C₈H₆BrN
MW: 196.05 g/mol
Boiling Pt: 78…82 °C (0.3 mmHg)
Melting Pt: 55
MDL Number: MFCD00031538
CAS Number: 41963-20-6
UN: 3439
ADR: 6.1 III