Propidium Iodide, Sigma-Aldrich®
Supplier: Merck
Warning
Synonyms: 3,8-Diamino-5-[3-(diethylmethylammonio)propyl]-6-phenylphenanthridinium diiodide, PI, 3,8-Diamino-5-(3-(diethyl(methyl)ammonio)propyl)-6-phenylphenanthridin-5-ium iodide, 3,8-Diamino-5-(3-diethylaminopropyl)-6-phenylphenanthridinium iodide methiodide
M-Clarity™ quality level = MQ200
81845-100MG 81845-25MG
SIAL81845-100MGEA 188 EUR
SIAL81845-100MG SIAL81845-25MG
Propidium Iodide, Sigma-Aldrich®
Propidium iodide
Formula: C₂₇H₃₄I₂N₄
MW: 668.4 g/mol
MDL Number: MFCD00011921
CAS Number: 25535-16-4
EINECS: 247-081-0