p-Toluidinium chloride, Sigma-Aldrich®
Supplier: Merck
Danger
M-Clarity™ quality level = MQ100
89650-100G 89650-500G
SIAL89650-100GEA 82.6 EUR
SIAL89650-100G SIAL89650-500G
p-Toluidinium chloride, Sigma-Aldrich®
p-Toluidinium chloride
Formula: C₇H₁₀ClN
Melting Pt: 239…242 °C
Density: 1.19 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00035497
CAS Number: 540-23-8
EINECS: 208-740-8
UN: 2811
ADR: 6.1,II