1-(Trimethylsilyl)imidazole, Sigma-Aldrich®
Supplier: Merck
Synonyms: TSIM, N-(Trimethylsilyl)imidazole, N-(Trimethylsilyl)-1H-imidazole, TMSI, 1-(Trimethylsilyl)-1H-imidazole
M-Clarity™ quality level = MQ200
92751-250ML 92751-50ML
SIAL92751-250MLEA 399 EUR
SIAL92751-250ML SIAL92751-50ML
1-(Trimethylsilyl)imidazole, Sigma-Aldrich®
1-(Trimethylsilyl)imidazole
Formula: C₆H₁₂N₂Si
Boiling Pt: 209 °C (1013 hPa)
Melting Pt: –42 °C
Density: 0.95 g/cm³ (20 °C)
Flash Pt: 61 °C
MDL Number: MFCD00005280
CAS Number: 18156-74-6
EINECS: 242-040-3