Acrolein diethyl acetal, Sigma-Aldrich®

Supplier: Merck

Synonyms: Acrolein Acetal, 1,1-Diethoxy-2-propene, 3,3-Diethoxy-1-propene

M-Clarity™ quality level = MQ200

A24001-100G A24001-25G
SIALA24001-100GEA 172 EUR
SIALA24001-100G SIALA24001-25G
Acrolein diethyl acetal, Sigma-Aldrich®
Acrolein diethyl acetal
Formula: C₇H₁₄O₂
Boiling Pt: 125 °C (1013 hPa)
Melting Pt: –77 °C
Density: 0.8543 g/cm³ (15 °C)
MDL Number: MFCD00009239
CAS Number: 3054-95-3
EINECS: 221-276-0
UN: 2374
ADR: 3,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR