2-Aminoanthracene, Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ100
A38800-1G A38800-25G A38800-5G
SIALA38800-1GEA 41.4 EUR
SIALA38800-1G SIALA38800-25G SIALA38800-5G
2-Aminoanthracene, Sigma-Aldrich®
2-Aminoanthracene
Formula: C₁₄H₁₁N
Boiling Pt: 371 °C (1013 hPa)
Melting Pt: 218…219 °C
Density: >1 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003582
CAS Number: 613-13-8
EINECS: 210-330-9