2-(Aminomethyl)pyridine, Sigma-Aldrich®
Supplier: Merck
Synonyms: 2-Pyridylmethylamine, 2-Picolylamine, Pyridin-2-ylmethanamine, (2-Pyridyl)methylamine, 2-Picolamine
M-Clarity™ quality level = MQ100
A65204-25G A65204-100G A65204-5G
SIALA65204-25GEA 82.4 EUR
SIALA65204-25G SIALA65204-100G SIALA65204-5G
2-(Aminomethyl)pyridine, Sigma-Aldrich®
2-(Aminomethyl)pyridine
Formula: C₆H₈N₂
MW: 108.14 g/mol
Boiling Pt: 197
Melting Pt: 81
Density: 1.0583 g/cm³ (20 °C)
MDL Number: MFCD00006360
CAS Number: 3731-51-9
EINECS: 223-090-5