2-Amino-4-methylthiazole, Sigma-Aldrich®
Supplier: Merck
Synonyms: 4-Methylthiazol-2-ylamine, 4-Methylthiazol-2-amine, 2-Amino-4-methylthiazol
M-Clarity™ quality level = MQ100
A66006-25G A66006-5G
SIALA66006-25GEA 76 EUR
SIALA66006-25G SIALA66006-5G
2-Amino-4-methylthiazole, Sigma-Aldrich®
2-Amino-4-methylthiazole
Formula: C₄H₆N₂S
MW: 114.17 g/mol
Boiling Pt: 231
Melting Pt: 42
Density: 1.202 g/cm³ (50 °C)
MDL Number: MFCD00005329
CAS Number: 1603-91-4
EINECS: 216-505-6
Merck Index: 13,00449