2-Amino-2-methyl-1,3-propanediol, Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ300
A9754-100G A9754-25G A9754-500G
SIALA9754-100GEA 128 EUR
SIALA9754-100G SIALA9754-25G SIALA9754-500G
2-Amino-2-methyl-1,3-propanediol, Sigma-Aldrich®
2-Amino-2-methyl-1,3-propanediol
Formula: C₄H₁₁NO₂
Boiling Pt: 272 °C (1013 hPa)
Melting Pt: 102…105 °C
Density: 1.211 g/cm³ (20 °C)
MDL Number: MFCD00004678
CAS Number: 115-69-5
EINECS: 204-100-7
Merck Index: 13,00447