Benzyl mercaptan, Sigma-Aldrich®
Supplier: Merck
Synonyms: Toluene-α-thiol, α-Toluenethiol
M-Clarity™ quality level = MQ100
B25401-100G B25401-5G
SIALB25401-100GEA 48.6 EUR
SIALB25401-100G SIALB25401-5G
Benzyl mercaptan, Sigma-Aldrich®
Benzyl mercaptan
Formula: C₇H₈S
MW: 124.21 g/mol
Boiling Pt: 195
Melting Pt: 29
Density: 1.055 g/cm³ (20 °C)
Flash Pt: 70
Storage Temperature: Ambient
MDL Number: MFCD00004867
CAS Number: 100-53-8
EINECS: 202-862-5
UN: 2810
ADR: 6.1 II
Merck Index: 12,09456