2-Bromethylamine hydrobromide, Sigma-Aldrich®
Supplier: Merck
Warning
Synonyms: 2-Bromoethylammonium bromide, 2-Bromoethylamine hydrobromide
M-Clarity™ quality level = MQ200
B65705-100G B65705-25G
SIALB65705-100GEA 98.1 EUR
SIALB65705-100G SIALB65705-25G
2-Bromethylamine hydrobromide, Sigma-Aldrich®
2-Bromethylamine hydrobromide
Formula: BrCH₂CH₂NH₃Br
MW: 204.89 g/mol
Melting Pt: 170…173 °C
Storage Temperature: Ambient
MDL Number: MFCD00012886
CAS Number: 2576-47-8
EINECS: 219-924-2