N-(2-Bromoethyl)phthalimide, Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ200
B66302-100G
B66302-25G
SIALB66302-100GEA
78.3
EUR
SIALB66302-100G
SIALB66302-25G
N-(2-Bromoethyl)phthalimide, Sigma-Aldrich®
N-(2-Bromoethyl)phthalimide
Formula:
C₁₀H₈BrNO₂ Boiling Pt: 318 °C (1013 hPa) Melting Pt: 79…82 °C |
MDL Number:
MFCD00005902 CAS Number: 574-98-1 EINECS: 209-379-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...