Corn oil, Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ100
C8267-2.5L
C8267-500ML
SIALC8267-2.5LEA
205
EUR
SIALC8267-2.5L
SIALC8267-500ML
Corn oil, Sigma-Aldrich®
Corn oil
Melting Pt:
–10 °C Density: 0.918 g/cm³ (20 °C) |
MDL Number:
MFCD00130865 CAS Number: 8001-30-7 EINECS: 232-281-2 |