Oil of castor, Sigma-Aldrich®

Supplier: Merck

Synonyms: Castor oil, Ricinus oil, Oleum Ricini s. Castoris, Oil of Palma Christi

M-Clarity™ quality level = MQ200

C9606-1L C9606-500ML
SIALC9606-1LEA 429 EUR
SIALC9606-1L SIALC9606-500ML
Oil of castor, Sigma-Aldrich®
Oil of castor
Boiling Pt: 313 °C (1013 hPa)
Melting Pt: –10 °C
Density: 0.95 g/cm³ (20 °C)
Flash Pt: >113 °C (closed cup)
MDL Number: MFCD00130746
CAS Number: 8001-79-4
EINECS: 232-293-8
Merck Index: 13,01908

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR