1,4-Dibromo-2-nitrobenzene, Sigma-Aldrich®
Supplier: Merck
Warning
Synonyms: 2,5-Dibromonitrobenzene
M-Clarity™ quality level = MQ100
D42003-100G D42003-25G
SIALD42003-100GEA 121 EUR
SIALD42003-100G SIALD42003-25G
1,4-Dibromo-2-nitrobenzene, Sigma-Aldrich®
1,4-Dibromo-2-nitrobenzene
Formula: C₆H₃Br₂NO₂
MW: 280.9 g/mol
Melting Pt: 81…85 °C
Density: 2.374 g/cm³ (20 °C)
MDL Number: MFCD00007046
CAS Number: 3460-18-2
EINECS: 222-404-8