Methyl chloroformate, Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ200
M35304-100G
M35304-500G
M35304-5G
SIALM35304-100GEA
51.5
EUR
SIALM35304-100G
SIALM35304-500G
SIALM35304-5G
Methyl chloroformate, Sigma-Aldrich®
Methyl chloroformate
Formula:
C₂H₃ClO₂ MW: 94.5 g/mol Boiling Pt: 71…72 °C (1013 hPa) Melting Pt: –61 °C Density: 1.23 g/cm³ (20 °C) Flash Pt: 45 |
MDL Number:
MFCD00000639 CAS Number: 79-22-1 EINECS: 201-187-3 UN: 1238 ADR: 6.1 I Merck Index: 12,06123 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...