Methyl methacrylate, Sigma-Aldrich®
Supplier: Merck
Danger
Synonyms: Methacrylic acid methyl ester
M-Clarity™ quality level = MQ200
M55909-10L M55909-1L M55909-25L M55909-25ML M55909-2L M55909-500ML M55909-17L
SIALM55909-10LEA 346 EUR
SIALM55909-10L SIALM55909-1L SIALM55909-25L SIALM55909-25ML SIALM55909-2L SIALM55909-500ML SIALM55909-17L
Methyl methacrylate, Sigma-Aldrich®
Methyl methacrylate (monomer)
Formula: CH₂=C(CH₃)COOCH₃
MW: 100.12 g/mol
Boiling Pt: 100
Melting Pt: –48 °C
Density: 0.93 g/cm³ (20 °C)
Flash Pt: 8
Storage Temperature: Ambient
MDL Number: MFCD00008587
CAS Number: 80-62-6
EINECS: 201-297-1
UN: 1247
ADR: 3 II
REACH: 01-2119452498-28
Merck Index: 12,06005