Dimethyl sulphone, Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ200
M81705-100G M81705-500G M81705-5G
SIALM81705-100GEA 43.7 EUR
SIALM81705-100G SIALM81705-500G SIALM81705-5G
Dimethyl sulphone, Sigma-Aldrich®
Dimethyl sulphone
Formula: C₂H₆O₂S
Boiling Pt: 238 °C (1013 hPa)
Melting Pt: 107…110 °C
Density: 1.17 g/cm³ (110 °C)
MDL Number: MFCD00007566
CAS Number: 67-71-0
EINECS: 200-665-9
Merck Index: 13,03284