7-Nitro-1H-indazole, Sigma-Aldrich®
Supplier: Merck
Synonyms: 7-Nitroindazole, 7-NINA, 7-Nitro-1H-indazole
M-Clarity™ quality level = MQ100
N7778-1G N7778-5G
SIALN7778-1GEA 169 EUR
SIALN7778-1G SIALN7778-5G
7-Nitro-1H-indazole, Sigma-Aldrich®
7-Nitro-1H-indazole
Formula: C₇H₅N₃O₂
MW: 163.14 g/mol
Melting Pt: 190
Density: 1.56 g/cm³ (293 °C)
MDL Number: MFCD00022789
CAS Number: 2942-42-9
EINECS: 220-934-4