Sodium oleate, Sigma-Aldrich®
Supplier: Merck
Synonyms: Oleic acid sodium salt, 9Z-Octadecenoic Acid, C18:1(9Z), cis-9-Octadecenoic Acid
M-Clarity™ quality level = MQ200
O7501-10G O7501-1G O7501-250MG O7501-5G
SIALO7501-10GEA 1020 EUR
SIALO7501-10G SIALO7501-1G SIALO7501-250MG SIALO7501-5G
Sodium oleate, Sigma-Aldrich®
Sodium oleate
Formula: C₁₈H₃₃NaO₂
MW: 304.45 g/mol
Melting Pt: 232…235 °C
Density: 1.02 g/cm³ (20 °C)
Storage Temperature: Freezer
MDL Number: MFCD00004438
CAS Number: 143-19-1
EINECS: 205-591-0