Phthalaldehyde, Sigma-Aldrich®
Supplier: Merck
|
Danger
|
Synonyms:
1,2-Phthalic dicarboxaldehyde, Phthaldialdehyde, o-Phthalaldehyde, OPA, Phthaldicarboxaldehyde, Benzene-1,2-dicarboxaldehyde
M-Clarity™ quality level = MQ200
P1378-100G
P1378-1G
P1378-25G
P1378-5G
SIALP1378-100GEA
843
EUR
SIALP1378-100G
SIALP1378-1G
SIALP1378-25G
SIALP1378-5G
Phthalaldehyde, Sigma-Aldrich®
Phthalaldehyde
Formula:
C₈H₆O₂ Boiling Pt: 266 °C (1013 hPa) Melting Pt: 54…56 °C Density: 1.13 g/cm³ (20 °C) Flash Pt: 132 °C |
MDL Number:
MFCD00003335 CAS Number: 643-79-8 EINECS: 211-402-2 UN: 2923 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...