p-Phenetidine, Sigma-Aldrich®
Supplier: Merck
Warning
M-Clarity™ quality level = MQ200
P14815-100G P14815-500G
SIALP14815-100GEA 50.5 EUR
SIALP14815-100G SIALP14815-500G
p-Phenetidine, Sigma-Aldrich®
p-Phenetidine
Formula: C₈H₁₁NO
Boiling Pt: 254 °C (1013 hPa)
Melting Pt: 4 °C
Density: 1.06 g/cm³ (20 °C)
Flash Pt: 120 °C
MDL Number: MFCD00007865
CAS Number: 156-43-4
EINECS: 205-855-5
UN: 2311
ADR: 6.1,III
Merck Index: 12,07373