Pentaerythritol, Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ200
P4755-1KG-A
SIALP4755-1KG-AEA 58.5 EUR
SIALP4755-1KG-A
Pentaerythritol, Sigma-Aldrich®
Pentaerythritol
Formula: C₅H₁₂O₄
Boiling Pt: 380 °C (1013 hPa)
Melting Pt: 260…264 °C
Density: 1.511 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00004692
CAS Number: 115-77-5
EINECS: 204-104-9
Merck Index: 12,07245