3-Pyridylmethanol, Sigma-Aldrich®
Supplier: Merck
|
Warning
|
Synonyms:
3-(Hydroxymethyl)pyridine, 3-Pyridinemethanol, 3-Pyridylcarbinol, Pyridine-3-methanol, 3-Pyridylmethanol
M-Clarity™ quality level = MQ200
P66807-100G
P66807-5G
SIALP66807-100GEA
107
EUR
SIALP66807-100G
SIALP66807-5G
3-Pyridylmethanol, Sigma-Aldrich®
3-Pyridylmethanol
Formula:
C₆H₇NO Boiling Pt: 266 °C (1013 hPa) Melting Pt: –7 °C Density: 1.1357 g/cm³ (20 °C) |
MDL Number:
MFCD00006407 CAS Number: 100-55-0 EINECS: 202-864-6 Merck Index: 12,06614 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...