Safranine O, Sigma-Aldrich®
Supplier: SIGMA ALDRICH MICROSCOPY
Synonyms: Gossypimine, Safranin O, Safranin Y, Safranin A, 3,7-Diamino-2,8-dimethyl-5-phenylphenazinium chloride, 3,7-Diamino-2,8-dimethyl-5-phenylphenazin-5-ium chloride, Safranin T, CI 50240, Basic Red 2, Safranin, Cotton Red
M-Clarity™ quality level = MQ200
S8884-100G S8884-25G
S8884-100GEA 406 EUR
S8884-100G S8884-25G
Safranine O, Sigma-Aldrich®
Safranine O
Formula: C₂₀H₁₉ClN₄
Density: 1.447 g/cm³ (22 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011759
CAS Number: 477-73-6
EINECS: 207-518-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3dc0c31be4a55937a0b5a174dbed0c0bc902b4a8&vid=0a2ca33d8ae9a3b4dc6b34fd80a483a5b5b6257a&prid=web&clid=SAPDAM