Thiophene-2-carbaldehyde, Sigma-Aldrich®
Supplier: Merck
Warning
Synonyms: 2-Thiophenecarboxaldehyde, Thiophene-2-carboxaldehyde
M-Clarity™ quality level = MQ200
T32409-100G T32409-25G
SIALT32409-100GEA 53.8 EUR
SIALT32409-100G SIALT32409-25G
Thiophene-2-carbaldehyde, Sigma-Aldrich®
Thiophene-2-carbaldehyde
Formula: C₅H₄OS
Boiling Pt: 198 °C (1013 hPa)
Melting Pt: ? 10 °C
Density: 1.2264 g/cm³ (20 °C)
Flash Pt: 78 °C
MDL Number: MFCD00005429
CAS Number: 98-03-3
EINECS: 202-629-8