Eucalyptol, Sigma-Aldrich®

Supplier: Merck
Danger

Synonyms: 1,8-Cineole, 1,3,3-Trimethyl-2-oxabicyclo[2.2.2]octane

M-Clarity™ quality level = MQ300

W246506-10KG-K W246506-1KG-K W246506-20KG-K
SIALW246506-10KG-KEA 1310 EUR
SIALW246506-10KG-K SIALW246506-1KG-K SIALW246506-20KG-K
Eucalyptol, Sigma-Aldrich®
Eucalyptol
Formula: C₁₀H₁₈O
MW: 154.25 g/mol
Boiling Pt: 176…177 °C (1013 hPa)
Melting Pt: 1.5
Density: 0.92 g/cm³ (20 °C)
Flash Pt: 46
Storage Temperature: Ambient
MDL Number: MFCD00167977
CAS Number: 470-82-6
EINECS: 207-431-5
UN: 3271
ADR: 3 III
Merck Index: 12,03940

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR