Furfuryl alcohol, Sigma-Aldrich®
Supplier: Merck
Danger
Synonyms: Furfurol
M-Clarity™ quality level = MQ100
W249106-1KG-K W249106-25KG-K W249106-5KG-K
SIALW249106-1KG-KEA 73.7 EUR
SIALW249106-1KG-K SIALW249106-25KG-K SIALW249106-5KG-K
Furfuryl alcohol, Sigma-Aldrich®
Furfuryl alcohol
Formula: C₅H₆O₂
Boiling Pt: 171 °C (1013 hPa)
Melting Pt: –31 °C
Density: 1.13 g/cm³ (20 °C)
Flash Pt: 75 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00003252
CAS Number: 98-00-0
EINECS: 202-626-1
UN: 2874
ADR: 6.1,III
Merck Index: 13,04326