(±)-Tetrahydrofurfuryl alcohol, Sigma-Aldrich®
Supplier: Merck
|
Warning
|
Synonyms:
(±)-Tetrahydro-2-furfuryl alcohol, (Tetrahydrofuran-2-yl)methanol, (±)-Tetrahydro-2-furyl methanol, (±)-THFA
M-Clarity™ quality level = MQ300
W305618-10KG-K
W305618-1KG-K
W305618-25KG-K
SIALW305618-10KG-KEA
303
EUR
SIALW305618-10KG-K
SIALW305618-1KG-K
SIALW305618-25KG-K
(±)-Tetrahydrofurfuryl alcohol, Sigma-Aldrich®
(±)-Tetrahydrofurfuryl alcohol
Formula:
C₅H₁₀O₂ Boiling Pt: 178 °C (1013 hPa) Melting Pt: < –80 °C Density: 1.0543 g/cm³ (20 °C) Flash Pt: 75 °C (closed cup) Storage Temperature: Ambient |
MDL Number:
MFCD00005372 CAS Number: 97-99-4 EINECS: 202-625-6 Merck Index: 12,09353 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...