2-Acetylthiazole, Sigma-Aldrich®
Supplier: Merck
Synonyms: 1-(Thiazol-2-yl)-1-ethanone
M-Clarity™ quality level = MQ300
W332801-100G-K W332801-1KG-K W332801-5KG-K
SIALW332801-100G-KEA 335 EUR
SIALW332801-100G-K SIALW332801-1KG-K SIALW332801-5KG-K
2-Acetylthiazole, Sigma-Aldrich®
2-Acetylthiazole
Formula: C₅H₅NOS
Boiling Pt: 213 °C (1013 hPa)
Melting Pt: 65.5 °C
Density: 1.22 g/cm³ (20 °C)
MDL Number: MFCD00005324
CAS Number: 24295-03-2
EINECS: 246-134-5