2-Ethyl-4-methylthiazole, Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ300
W368008-100G-K W368008-1KG-K W368008-25G-K
SIALW368008-100G-KEA 322 EUR
SIALW368008-100G-K SIALW368008-1KG-K SIALW368008-25G-K
2-Ethyl-4-methylthiazole, Sigma-Aldrich®
2-Ethyl-4-methylthiazole
Formula: C₆H₉NS
MW: 127.21 g/mol
Boiling Pt: 161
Density: 1.0231 g/cm³ (25 °C)
MDL Number: MFCD00036621
CAS Number: 15679-12-6
EINECS: 239-757-9