Glutaronitrile ≥99%
Supplier: Thermo Fisher Scientific
|
Warning
|
Purity: 99%
CAS No.: 544-13-8
Molecular Formula: C5H6N2
Formula Weight: 94.12
Flash Point: 112°(234°F)
Refractive Index: 1.4350
Melting Point: -29°
Boiling Point: 286-288°
Beilstein Reference: 1738385
Merck Reference: 14,4474
UN No.: 3276
MDL No.: MFCD00001970
Formula:
C₅H₆N₂ Boiling Pt: 286…288 °C Melting Pt: –29 °C Density: 0.990 Flash Pt: 112 °C (234 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00001970 CAS Number: 544-13-8 EINECS: 208-861-6 UN: 3276 ADR: 6.1,III Merck Index: 14,4474 |