3-Diethylaminopropylamine, Sigma-Aldrich®
Supplier: Merck
Synonyms:
3-Diethylamino-1-propylamine, DEAPA, N,N-Diethyl-1,3-propanediamine, N,N-Diethyl-1,3-diaminopropane, 3-Aminopropyldiethylamine, 3-(Diethylamino)propylamine
M-Clarity™ quality level = MQ200
549975-2L
549975-100ML
549975-500ML
SIAL549975-2LEA
190
EUR
SIAL549975-2L
SIAL549975-100ML
SIAL549975-500ML
3-Diethylaminopropylamine, Sigma-Aldrich®
3-Diethylaminopropylamine
Formula:
C₇H₁₈N₂ Boiling Pt: 168…171 °C (1013 hPa) Melting Pt: –60 °C Density: 0.8283 g/cm³ (20 °C) Flash Pt: 51 °C |
MDL Number:
MFCD00008218 CAS Number: 104-78-9 EINECS: 203-236-4 UN: 2684 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...