tert-Butanol-[D1], Sigma-Aldrich®

Supplier: Merck
Danger

Synonyms: tert-Butyl alcohol-D1, Trimethylcarbinol-D1, 2-Methyl-2-propanol-D1

M-Clarity™ quality level = MQ200

175765-25G 175765-5G
SIAL175765-25GEA 209 EUR
SIAL175765-25G SIAL175765-5G
tert-Butanol-[D1], Sigma-Aldrich®
tert-Butanol-[D1]
Formula: C₄H₉DO
Density: 0.797 g/cm³ (20 °C)
Storage Temperature: Ambient
CAS Number: 3972-25-6
EINECS: 223-597-1
UN: 1120
ADR: 3,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR