3-(Bromomethyl)phenylboronic acid, Sigma-Aldrich®

Supplier: Merck


M-Clarity™ quality level = MQ100

679445-1G 679445-5G
SIAL679445-1GEA 62.3 EUR
SIAL679445-1G SIAL679445-5G
3-(Bromomethyl)phenylboronic acid, Sigma-Aldrich®
3-(Bromomethyl)phenylboronic acid
Formula: C₇H₈BBrO₂
MDL Number: MFCD01632207
CAS Number: 51323-43-4
UN: 3261
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR