(±)-3-Butene-1,2-diol, Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ100
488216-100ML 488216-25ML
SIAL488216-100MLEA 789 EUR
SIAL488216-100ML SIAL488216-25ML
(±)-3-Butene-1,2-diol, Sigma-Aldrich®
(±)-3-Butene-1,2-diol
Formula: C₄H₈O₂
Boiling Pt: 95…96 °C (1013 hPa)
Density: 1.047 g/cm³ (20 °C)
MDL Number: MFCD00239410
CAS Number: 497-06-3
EINECS: 207-835-1