(±)-γ-Heptalactone, Sigma-Aldrich®
Supplier: Merck
Synonyms:
5-Propyldihydrofuran-2(3H)-one, gamma-Enanthonolactone, gamma-Heptalactone, gamma-Heptanolactone, (±)-Heptan-4-olide
M-Clarity™ quality level = MQ300
W253901-SAMPLE
W253901-100G-K
W253901-1KG-K
W253901-5KG-K
SIALW253901-SAMPLEEA
54.3
EUR
SIALW253901-SAMPLE
SIALW253901-100G-K
SIALW253901-1KG-K
SIALW253901-5KG-K
(±)-γ-Heptalactone, Sigma-Aldrich®
(±)-γ-Heptalactone
Formula:
C₇H₁₂O₂ |
MDL Number:
MFCD00036498 CAS Number: 105-21-5 EINECS: 203-279-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...