(±)-γ-Nonalactone, Sigma-Aldrich®

Supplier: Merck

Synonyms: gamma-Nonalactone, (±)-Nonan-4-olide, gamma-Nonanolactone, 5-Pentyldihydrofuran-2(3H)-one, gamma-Pelargonolactone

M-Clarity™ quality level = MQ300

W278106-10KG-K W278106-1KG-K W278106-25KG-K
SIALW278106-10KG-KEA 427 EUR
SIALW278106-10KG-K SIALW278106-1KG-K SIALW278106-25KG-K
(±)-γ-Nonalactone, Sigma-Aldrich®
(±)-γ-Nonalactone
Formula: C₉H₁₆O₂
Density: 0.97 g/cm³ (20 °C)
MDL Number: MFCD00005403
CAS Number: 104-61-0
EINECS: 203-219-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR