(±)-γ-Undecalactone, Sigma-Aldrich®

Supplier: Merck

Synonyms: (±)-Undecan-4-olide, Gamma-undecalactone, Undecanoic gamma-Lactone

M-Clarity™ quality level = MQ300

W309109-10KG-K W309109-1KG-K W309109-5KG-K
SIALW309109-10KG-KEA 527 EUR
SIALW309109-10KG-K SIALW309109-1KG-K SIALW309109-5KG-K
(±)-γ-Undecalactone, Sigma-Aldrich®
(±)-γ-Undecalactone
Formula: C₁₁H₂₀O₂
Boiling Pt: 297 °C (1013 hPa)
Density: 0.9414 g/cm³ (20 °C)
MDL Number: MFCD00005405
CAS Number: 104-67-6
EINECS: 203-225-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR